Anti-IgG2b Mouse Antibody (Alexa Fluor® 555) [clone: 2B10A8]

Supplier: Southern Biotechnology
3070-32
103264-848EA 377.92 USD
103264-848
Anti-IgG2b Mouse Antibody (Alexa Fluor® 555) [clone: 2B10A8]
Antibodies
Alexa Fluor 555 Anti-Rat IgG2b antibody for use in immunohistochemistry and western blotting assays.

Type: Secondary
Antigen: IgG2b
Clonality: Monoclonal
Clone: 2B10A8
Conjugation: Alexa Fluor® 555
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR