Anti-IgG2a Mouse Antibody (Alexa Fluor® 555) [clone: 2A8F4]

Supplier: Southern Biotechnology
3065-32
103264-842EA 377.92 USD
103264-842
Anti-IgG2a Mouse Antibody (Alexa Fluor® 555) [clone: 2A8F4]
Antibodies
Alexa Fluor 555 Anti-Rat IgG2a antibody for use in immunohistochemistry assays.

Type: Secondary
Antigen: IgG2a
Clonality: Monoclonal
Clone: 2A8F4
Conjugation: Alexa Fluor® 555
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR