Anti-IgG1 Mouse Antibody (Alexa Fluor® 488) [clone: G11C5]

Supplier: Southern Biotechnology
3061-30
103264-832EA 377.92 USD
103264-832
Anti-IgG1 Mouse Antibody (Alexa Fluor® 488) [clone: G11C5]
Antibodies
Alexa Fluor 488 Anti-Rat IgG1 antibody for use in flow cytometry and immunohistochemistry assays.

Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone: G11C5
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR