Anti-IgG (H+L) Goat Antibody (Biotin)

Supplier: Southern Biotechnology
3051-08
103264-826EA 187 USD
103264-826
Anti-IgG (H+L) Goat Antibody (Biotin)
Antibodies
Goat Anti-Rat IgG-Biotin antibody with minimal reactivity to mouse and human proteins for use in ELISA and flow cytometry assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR