Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
3032-05
103264-814EA 252.38 USD
103264-814
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat F(ab')2 Anti-Rat IgG-HRP antibody for use in ELISA assays.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR