Anti-IgG Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
3030-09
100244-488EA 309.92 USD
100244-488
Anti-IgG Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat Anti-Rat IgG-PE antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR