Anti-IgM Goat Antibody (Alexa Fluor® 488)

Supplier: Southern Biotechnology
3020-30
103264-810EA 187 USD
103264-810
Anti-IgM Goat Antibody (Alexa Fluor® 488)
Antibodies
Goat Anti-Rat IgM-Alexa Fluor 488 antibody for use in flow cytometry, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR