Anti-Ig Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
3000-05
103264-808EA 215.77 USD
103264-808
Anti-Ig Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Fab Anti-Rat Ig-HRP antibody with minimal reactivity to mouse proteins for use in ELISA assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR