Anti-Lambda Goat Antibody (Alexa Fluor® 647)

Supplier: Southern Biotechnology
2072-31
103260-030EA 330.85 USD
103260-030
Anti-Lambda Goat Antibody (Alexa Fluor® 647)
Antibodies
Goat F(ab')2 Anti-Human Lambda-Alexa Fluor 647 antibody for use in flow cytometry and western blotting assays.

Type: Secondary
Antigen: Lambda
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR