Anti-Lambda Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
2071-05
103260-024EA 338.69 USD
103260-024
Anti-Lambda Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Human Lambda-HRP antibody with minimal reactivity to mouse proteins for use in ELISA assays.

Type: Secondary
Antigen: Lambda
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR