Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Human Lambda Antibody (Alexa Fluor® 555)

Supplier: SOUTHERN BIOTECHNOLOGY MS
2070-32
103260-016
Goat Anti-Human Lambda Antibody (Alexa Fluor® 555)
Goat Anti-Human Lambda-Alexa Fluor 555 antibody for use in immunohistochemistry / immunocytochemistry and western blotting assays.

Type: Secondary
Antigen: Lambda
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty