Anti-Lambda Goat Antibody

Supplier: Southern Biotechnology
2070-01
100244-370EA 158.23 USD
100244-370
Anti-Lambda Goat Antibody
Antibodies
Purified Goat Anti-Human Lambda antibody for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, immunoprecipitation, ELISpot, and stimulation assays and for immobilization in surface plasmon resonance.

Type: Secondary
Antigen: Lambda
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR