Anti-Kappa Goat Antibody (Alexa Fluor® 488)

Supplier: Southern Biotechnology
2062-30
103260-006EA 330.85 USD
103260-006
Anti-Kappa Goat Antibody (Alexa Fluor® 488)
Antibodies
Goat F(ab')2 Anti-Human Kappa-Alexa Fluor 488 antibody for use in flow cytometry, immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: Kappa
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR