Anti-Kappa Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
2061-09
103260-004EA 525.69 USD
103260-004
Anti-Kappa Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat Anti-Human Kappa-PE antibody with minimal reactivity to mouse proteins for use in flow cytometry assays.

Type: Secondary
Antigen: Kappa
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR