Anti-IgG Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
2049-02
103264-784EA 468.15 USD
103264-784
Anti-IgG Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat Anti-Human IgG-FITC antibody with minimal reactivity to rhesus and cynomolgus proteins for use in fluorescent assays.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR