Anti-IgG F(c) Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
2048-05
103265-656EA 143.85 USD
103265-656
Anti-IgG F(c) Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Human IgG Fc-HRP antibody for use in ELISA and western blotting assays.

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR