Anti-IgG F(c) Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
2048-02
103265-652EA 94.15 USD
103265-652
Anti-IgG F(c) Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat Anti-Human IgG Fc-FITC antibody for use in western blotting and for other fluorescent assays.

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR