Anti-IgG F(c) Goat Antibody

Supplier: Southern Biotechnology
2048-01
103265-650EA 71.92 USD
103265-650
Anti-IgG F(c) Goat Antibody
Antibodies
Purified Goat Anti-Human IgG Fc antibody for use in ELISA and western blotting assays and for immobilization in surface plasmon resonance.

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR