Anti-Ig F(c) Goat Antibody

Supplier: Southern Biotechnology
2047-01
103265-644EA 100.69 USD
103265-644
Anti-Ig F(c) Goat Antibody
Antibodies
Purified Goat Anti-Human Ig Fc antibody for use in ELISA assays.

Type: Secondary
Antigen: Ig F(c)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR