Anti-IgG Fd Goat Antibody (AP (Alkaline Phosphatase))

Supplier: Southern Biotechnology
2046-04
103265-640EA 316.46 USD
103265-640
Anti-IgG Fd Goat Antibody (AP (Alkaline Phosphatase))
Antibodies
Goat Anti-Human IgG Fd-AP antibody for use in ELISA assays.

Type: Secondary
Antigen: IgG Fd
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR