Anti-IgG Fd Goat Antibody

Supplier: Southern Biotechnology
2046-01
103265-636EA 172.62 USD
103265-636
Anti-IgG Fd Goat Antibody
Antibodies
Purified Goat Anti-Human IgG Fd antibody for use in ELISA assays.

Type: Secondary
Antigen: IgG Fd
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR