Anti-IgM Goat Antibody

Supplier: Southern Biotechnology
2023-01
103264-752EA 137.31 USD
103264-752
Anti-IgM Goat Antibody
Antibodies
Purified Goat Anti-Human IgM antibody with minimal reactivity to mouse, bovine, and horse serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR