Anti-IgM Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
2022-09
100244-232EA 388.38 USD
100244-232
Anti-IgM Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat F(ab')2 Anti-Human IgM-PE antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR