Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Human IgM Antibody (HRP (Horseradish Peroxidase))

Supplier: SOUTHERN BIOTECHNOLOGY MS

2022-05
103264-744EA 163.46 USD InStock
103264-744
Goat Anti-Human IgM Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat F(ab')2 Anti-Human IgM-HRP antibody for use in ELISA, immunocytochemistry, and ELISpot assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty