Anti-IgM Goat Antibody (Alexa Fluor® 555)

Supplier: Southern Biotechnology
2020-32
103264-742EA 172.62 USD
103264-742
Anti-IgM Goat Antibody (Alexa Fluor® 555)
Antibodies
Goat Anti-Human IgM-Alexa Fluor 555 antibody for use in immunohistochemistry / immunocytochemistry and western blotting assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR