Anti-IgM Goat Antibody (Biotin)

Supplier: Southern Biotechnology
2020-08
100244-222EA 122.92 USD
100244-222
Anti-IgM Goat Antibody (Biotin)
Antibodies
Goat Anti-Human IgM-Biotin antibody for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, immunoprecipitation, ELISpot, and multiplex assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR