Anti-IgM Goat Antibody

Supplier: Southern Biotechnology
2020-01
100244-210EA 100.69 USD
100244-210
Anti-IgM Goat Antibody
Antibodies
Purified Goat Anti-Human IgM antibody for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, immunoprecipitation, ELISpot, multiplex, stimulation, purification, and depletion assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR