Anti-IgG F(c) Goat Antibody (Alexa Fluor® 555)

Supplier: Southern Biotechnology
2014-32
103264-720EA 302.08 USD
103264-720
Anti-IgG F(c) Goat Antibody (Alexa Fluor® 555)
Antibodies
Goat Anti-Human IgG Fc-Alexa Fluor 555 antibody with minimal reactivity to rabbit, mouse, rat, bovine, horse, hamster, goat, sheep, chicken, and guinea pig serum proteins for use in fluorescent assays.

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR