Anti-IgG F(c) Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
2014-09
103264-716EA 353.08 USD
103264-716
Anti-IgG F(c) Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat Anti-Human IgG Fc-PE antibody with minimal reactivity to rabbit, mouse, rat, bovine, horse, hamster, goat, sheep, chicken, and guinea pig serum proteins for use in flow cytometry and other fluorescent assays.

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR