Anti-IgG F(c) Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
2014-05
103264-712EA 230.15 USD
103264-712
Anti-IgG F(c) Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Human IgG Fc-HRP antibody with minimal reactivity to rabbit, mouse, rat, bovine, horse, hamster, goat, sheep, chicken, and guinea pig serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR