Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
2012-02
100244-204EA 108.54 USD
100244-204
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat F(ab')2 Anti-Human Ig-FITC antibody for use in flow cytometry and immunohistochemistry assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR