Anti-Ig Goat Antibody (Texas Red®)

Supplier: Southern Biotechnology
2010-07
100244-196EA 100.69 USD
100244-196
Anti-Ig Goat Antibody (Texas Red®)
Antibodies
Goat Anti-Human Ig-Texas Red antibody for use in flow cytometry and immunocytochemistry assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR