Anti-Ig Goat Antibody (bGal (beta Galactosidase))

Supplier: Southern Biotechnology
2010-06
100244-194EA 137.31 USD
100244-194
Anti-Ig Goat Antibody (bGal (beta Galactosidase))
Antibodies
Goat Anti-Human Ig-β-galactosidase antibody for use in ELISA and immunocytochemistry assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: bGal (beta Galactosidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR