Anti-IgG1 Mouse Antibody (RPE (R-Phycoerythrin)) [clone: SB139a]

Supplier: Southern Biotechnology
1940-09
103264-698EA 353.08 USD
103264-698
Anti-IgG1 Mouse Antibody (RPE (R-Phycoerythrin)) [clone: SB139a]
Antibodies
PE Anti-Hamster IgG1 antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone: SB139a
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR