Anti-IgG1 Mouse Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SB139a]

Supplier: Southern Biotechnology
1940-02
103264-692EA 266.77 USD
103264-692
Anti-IgG1 Mouse Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SB139a]
Antibodies
FITC Anti-Hamster IgG1 antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone: SB139a
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR