Anti-IgG3 Rat Antibody (RPE (R-Phycoerythrin)) [clone: SB76b]

Supplier: Southern Biotechnology
1191-09L 1191-09
103259-874EA 266.77 USD
103259-874 103259-876
Anti-IgG3 Rat Antibody (RPE (R-Phycoerythrin)) [clone: SB76b]
Antibodies
PE Anti-Mouse IgG3 antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgG3
Clonality: Monoclonal
Clone: SB76b
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR