Anti-IgG2b Rat Antibody (Biotin) [clone: SB74g]

Supplier: Southern Biotechnology
1186-08
103259-858EA 273.31 USD
103259-858
Anti-IgG2b Rat Antibody (Biotin) [clone: SB74g]
Antibodies
Biotin Anti-Mouse IgG2b antibody for use in ELISA, flow cytometry, immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG2b
Clonality: Monoclonal
Clone: SB74g
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR