Anti-Mouse Lambda Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: JC5-1]

Supplier: Southern Biotechnology
1175-02
100242-870EA 281.15 USD
100242-870
Anti-Mouse Lambda Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: JC5-1]
Antibodies
FITC Anti-Mouse Lambda antibody for use in flow cytometry, immunohistochemistry, and western blotting assays.

Type: Secondary
Antigen: Lambda
Clonality: Monoclonal
Clone: JC5-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR