Anti-Mouse Kappa Rat Antibody (Biotin) [clone: 187.1]

Supplier: Southern Biotechnology
1170-08
100242-862EA 266.77 USD
100242-862
Anti-Mouse Kappa Rat Antibody (Biotin) [clone: 187.1]
Antibodies
Biotin Anti-Mouse Kappa antibody for use in ELISA, flow cytometry, immunohistochemistry, western blotting, and immunoprecipitation assays.

Type: Secondary
Antigen: Kappa
Clonality: Monoclonal
Clone: 187.1
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR