Anti-IgG2a Rat Antibody (Alexa Fluor® 647) [clone: SB84a]

Supplier: Southern Biotechnology
1155-31
103259-846EA 281.15 USD
103259-846
Anti-IgG2a Rat Antibody (Alexa Fluor® 647) [clone: SB84a]
Antibodies
Alexa Fluor 647 Anti-Mouse IgG2a antibody for use in flow cytometry, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG2a
Clonality: Monoclonal
Clone: SB84a
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR