Anti-IgG1 Rat Antibody (Alexa Fluor® 555) [clone: SB77e]

Supplier: Southern Biotechnology
1144-32
103264-504EA 281.15 USD
103264-504
Anti-IgG1 Rat Antibody (Alexa Fluor® 555) [clone: SB77e]
Antibodies
Alexa Fluor 555 Anti-Mouse IgG1 antibody for use in immunohistochemistry / immunocytochemistry and western blotting assays.

Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone: SB77e
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR