Anti-IgG1 Rat Antibody [clone: SB77e]

Supplier: Southern Biotechnology
1144-01
103264-488EA 230.15 USD
103264-488
Anti-IgG1 Rat Antibody [clone: SB77e]
Antibodies
Purified Anti-Mouse IgG1 antibody for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone: SB77e
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR