Anti-IgM Rat Antibody [clone: SB73a]

Supplier: Southern Biotechnology
1142-01
103264-486EA 309.92 USD
103264-486
Anti-IgM Rat Antibody [clone: SB73a]
Antibodies
Purified F(ab')2 Anti-Mouse IgM antibody for use in ELISA, flow cytometry, and stimulation assays.

Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: SB73a
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR