Anti-IgM Rat Antibody (Alexa Fluor® 555) [clone: 1B4B1]

Supplier: Southern Biotechnology
1140-32
103264-484EA 281.15 USD
103264-484
Anti-IgM Rat Antibody (Alexa Fluor® 555) [clone: 1B4B1]
Antibodies
Alexa Fluor 555 Anti-Mouse IgM antibody for use in immunohistochemistry and western blotting assays.

Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: 1B4B1
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR