Anti-IgM Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1B4B1]

Supplier: Southern Biotechnology
1140-02
100242-796EA 281.15 USD
100242-796
Anti-IgM Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1B4B1]
Antibodies
FITC Anti-Mouse IgM antibody for use in flow cytometry, immunohistochemistry, and western blotting assays.

Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone: 1B4B1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR