Anti-IgE Rat Antibody (HRP (Horseradish Peroxidase)) [clone: 23G3]

Supplier: Southern Biotechnology
1130-05
100242-786EA 309.92 USD
100242-786
Anti-IgE Rat Antibody (HRP (Horseradish Peroxidase)) [clone: 23G3]
Antibodies
HRP Anti-Mouse IgE antibody for use in ELISA, immunohistochemistry, western blotting, and ELISpot assays.

Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone: 23G3
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR