Anti-IgD Rat Antibody (Pacific Blue™) [clone: 43430]

Supplier: Southern Biotechnology
1120-26
103264-472EA 286.38 USD
103264-472
Anti-IgD Rat Antibody (Pacific Blue™) [clone: 43430]
Antibodies
Pacific Blue Anti-Mouse IgD antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgD
Clonality: Monoclonal
Clone: 43430
Conjugation: Pacific Blue™
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR