Anti-IgD Rat Antibody [clone: 43430]

Supplier: Southern Biotechnology
1120-14
100242-778EA 425 USD
100242-778
Anti-IgD Rat Antibody [clone: 43430]
Antibodies
Low Endotoxin/Azide Free Anti-Mouse IgD antibody for use in flow cytometry, immunohistochemistry, immunoprecipitation, separation, costimulation, and proximity ligation assays.

Type: Secondary
Antigen: IgD
Clonality: Monoclonal
Clone: 43430
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR