Anti-IgE Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
1110-05
100242-764EA 258.92 USD
100242-764
Anti-IgE Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Mouse IgE-HRP antibody for use in ELISA, immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR