Anti-IgG3 Goat Antibody (AP (Alkaline Phosphatase))

Supplier: Southern Biotechnology
1103-04
103259-840EA 187 USD
103259-840
Anti-IgG3 Goat Antibody (AP (Alkaline Phosphatase))
Antibodies
Goat Anti-Mouse IgG3-AP antibody with minimal reactivity to human, bovine, and horse serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgG3
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR